Visual colour kits, nickel, MQuant®, Supelco®
Nikkeltest, 0,5-1-1,5-2-3-4-6-8-10 mg/l Ni, Dimethylglyoxime, MCD, 500 Tests
: Merck
Hurtige test, visuel farvesammenligning.
- Resultater på 5 minutter
- Brugervenlig
- Applikationstilpassede måleområder
- Bærbar
- Et udvalg af økonomiske refillpakker (RP) tilbydes
MQuant® with colour disc comparator
Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.
RP= Refil pakke
: Hvert testsæt indeholder alle de nødvendige reagenser, reaktionskuvetter, farvesammenligningskort eller sammenligningshjul og omfattende brugsvejledninger.
- Kemisk metode:Dimethylglyoxime
- Pakn:500 Tests
- Beskrivelse:Nikkeltest
- Model:MCD
- Område:0,5-1-1,5-2-3-4-6-8-10 mg/l Ni
- Cat. No.:1.14783.0001
- M-Clarity quality level:MQ100