Orders LinkContactUsLinkComponent
Accessories for empty columns HiScale™
Accessories for empty columns HiScale™
Accessory kit for HiScale™ 50, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
Katalog # 28-9663-75
Leverandør:   Cytiva
Accessories for empty columns HiScale™
Katalog # 28-9663-75
Leverandør:  Cytiva
Accessories for empty columns HiScale™
Accessories for empty columns HiScale™
Accessory kit for HiScale™ 50, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
Price
1.020,00 kr
Log ind for tilgængelighed
1 * 1 Each
Specifikationer
  • Beskrivelse:
    Accessory kit for HiScale™ 50, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
  • Seq:
    39