Accessories for empty columns HiScale™
Accessory kit for HiScale™ 26, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
Katalog # 28-9663-74
Leverandør: Cytiva
Gå til:
Product Details & Documents
Documents
Certifikater
Søg efter partinummerAffinity Purification using AKTA Start.pdf Anion Exchange using AKTA Start Desalting using AKTA Start
Manuel
AKTA Start Brochure
Produktlitteratur
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Yderligere produktdetaljer
Specifikationer
- Beskrivelse:Accessory kit for HiScale™ 26, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
- Seq:38