Centrifugerør, Axygen®
50 ml, Skruelåg, Sterile, 25 tubes and assembled caps per rack, 20 racks per case
525-1747
: Corning
PP, conical, printed graduation, with HDPE flat screw caps.
- Max. RCF: 12 000×g
- Easy one hand operation
- Large writing area for sample identification
- Lukkestørrelse:Skruelåg
- Pakket:25 tubes and assembled caps per rack, 20 racks per case
- Version:Sterile
- Kapacitet:50 ml
- Seq:08
- Cat. No.:525-1747
- Miljøcertificering:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Lav fremstillingspåvirkning