Orders LinkContactUsLinkComponent
Centrifugerør, Axygen®
50 ml, Skruelåg, Sterile, 25 tubes and assembled caps per rack, 20 racks per case
  525-1747
   Lav fremstillingspåvirkning
 :   Corning
Centrifugerør, Axygen®
  525-1747
 :  Corning
   Lav fremstillingspåvirkning
Centrifugerør, Axygen®
Centrifugerør, Axygen®
50 ml, Skruelåg, Sterile, 25 tubes and assembled caps per rack, 20 racks per case
1.450,00 kr
1 * 500 Items
PP, conical, printed graduation, with HDPE flat screw caps.
  • Max. RCF: 12 000×g
  • Easy one hand operation
  • Large writing area for sample identification
  • Lukkestørrelse:
    Skruelåg
  • Pakket:
    25 tubes and assembled caps per rack, 20 racks per case
  • Version:
    Sterile
  • Kapacitet:
    50 ml
  • Seq:
    08
  • Cat. No.:
    525-1747
  • Miljøcertificering:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Lav fremstillingspåvirkning