Cover glasses
High precision cover glasses, 18×18 mm, #1,5, Borosilikatglas
Catalog # 630-2185
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Cover slides suitable for fluorescence microscopy.
- Borosilicate glass D 263™ M of hydrolytic class 1
Recommended for objectives with high numerical aperture and resolution.
Documents
Certificates
Search by Lot NumberSpecifications
- Antal:#1,5
- Beskrivelse:High precision cover glasses
- L×B:18×18 mm
- Materiale:Borosilikatglas
- Seq:01