Orders LinkContactUsLinkComponent
Accessories for empty columns HiScale™
Tubing, outer Ø 2 mm, cut, for HiScale™ 50
Katalog # 28-9666-56
Leverandør:   Cytiva
Accessories for empty columns HiScale™
Katalog # 28-9666-56
Leverandør:  Cytiva
Accessories for empty columns HiScale™
Accessories for empty columns HiScale™
Tubing, outer Ø 2 mm, cut, for HiScale™ 50
Price
632,00 kr
Log ind for tilgængelighed
1 * 0,295 m
Specifikationer
  • Beskrivelse:
    Tubing, outer Ø 2 mm, cut, for HiScale™ 50
  • Seq:
    15