IV catheter, Introcan®
Product Details & Documents
Conventional IV catheter designed for single-handed insertion. Push-off plate on catheter hub facilitates single-handed puncture procedures. Sharp backcut bevel for minimal puncture trauma.
- Four fully embedded x-ray contrast lines
- Hydrophobic blood stoppers and lock fitting
- Colour-coded sizes
- Latex free and PVC free, catheter made of FEP
Meets ISO standard 10555-1/5 — same product properties as the Introcan Safety® (with PUR catheter) excluding the integrated, self-activating safety device for preventing needlestick injuries and the associated risk of infection.
Certifications: ISO 10555-1/5 — Same properties as the Introcan Safety® (with PUR catheter) excluding the integrated, self-activating safety device for preventing needlestick injuries and the associated risk of infection.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Ø×L:1,30×32 mm
- Måler:G18
- Farve:grøn
- Beskrivelse:IV catheter, Introcan®
- Seq:06