Orders LinkContactUsLinkComponent
IV catheter, Introcan®
IV catheter, Introcan®
IV catheter, Introcan®, grøn, G18, 1,30×32 mm
Katalog # BRAU4252322B
Leverandør:   B. Braun
IV catheter, Introcan®
Katalog # BRAU4252322B
Leverandør:  B. Braun
IV catheter, Introcan®
IV catheter, Introcan®
IV catheter, Introcan®, grøn, G18, 1,30×32 mm
Price
1.080,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents
Conventional IV catheter designed for single-handed insertion. Push-off plate on catheter hub facilitates single-handed puncture procedures. Sharp backcut bevel for minimal puncture trauma.
  • Four fully embedded x-ray contrast lines
  • Hydrophobic blood stoppers and lock fitting
  • Colour-coded sizes
  • Latex free and PVC free, catheter made of FEP
Meets ISO standard 10555-1/5 — same product properties as the Introcan Safety® (with PUR catheter) excluding the integrated, self-activating safety device for preventing needlestick injuries and the associated risk of infection.
Certifications: ISO 10555-1/5 — Same properties as the Introcan Safety® (with PUR catheter) excluding the integrated, self-activating safety device for preventing needlestick injuries and the associated risk of infection.
Documents
Specifikationer
  • Ø×L:
    1,30×32 mm
  • Måler:
    G18
  • Farve:
    grøn
  • Beskrivelse:
    IV catheter, Introcan®
  • Seq:
    06