Safety spectacles, SP1000™
Anti-fog, anti-scratch, sort, Clear PC
Katalog # 111-2161
Leverandør: HONEYWELL SAFETY
Gå til:
Product Details & Documents
These dual lens spectacles provide protection against impact, wind, dust and airborne debris. Ideal for dirty, high particulate work environments such as grinding, chipping, shaving and sanding.
- Soft and flexible nosepiece
- Soft, high quality face cushioning and padded temple tips
- Highly adjustable flame resistant headband
- Wrap-around eight-base dual lens style
- Top and bottom indirect ventilation and Dura-streme dual anti-fog/anti-scratch coating
Certifications: EN 166
Specifikationer
- Model:Anti-fog, anti-scratch
- Farve:sort
- Seq:01
- Linser:Clear PC