Orders LinkContactUsLinkComponent
Safety spectacles, SP1000™
Safety spectacles, SP1000™
Anti-fog, anti-scratch, sort, Clear PC
Katalog # 111-2161
Leverandør:   HONEYWELL SAFETY
Safety spectacles, SP1000™
Katalog # 111-2161
Leverandør:  HONEYWELL SAFETY
Safety spectacles, SP1000™
Safety spectacles, SP1000™
Anti-fog, anti-scratch, sort, Clear PC
Price
233,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These dual lens spectacles provide protection against impact, wind, dust and airborne debris. Ideal for dirty, high particulate work environments such as grinding, chipping, shaving and sanding.
  • Soft and flexible nosepiece
  • Soft, high quality face cushioning and padded temple tips
  • Highly adjustable flame resistant headband
  • Wrap-around eight-base dual lens style
  • Top and bottom indirect ventilation and Dura-streme dual anti-fog/anti-scratch coating
Certifications: EN 166
Specifikationer
  • Model:
    Anti-fog, anti-scratch
  • Farve:
    sort
  • Seq:
    01
  • Linser:
    Clear PC