Orders LinkContactUsLinkComponent
Anatomical Charts
Anatomical Charts
Chart, human inner, outer and middle ear, laminated, 500×670 mm
Katalog # 761-0471
Marketsource
Leverandør:   3B Scientific
Anatomical Charts
Katalog # 761-0471
Leverandør:  3B Scientific
Marketsource
Anatomical Charts
Chart, human inner, outer and middle ear, laminated, 500×670 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These colourful anatomical charts depict various parts of the human body.
  • Thick lamination ensures the chart does not curl up at the edges
  • UV treatment ensures the chart does not get a faded yellow colour over time
  • Printed on premium glossy UV resistant paper and thickly laminated on both sides
Leveringsinformation: Supplied with metal eyelets for easy hanging.
Specifikationer
  • Beskrivelse:
    Chart, human inner, outer and middle ear, laminated
  • B×H:
    500×670 mm