Anatomical Charts
Chart, human skeleton, laminated, 500×670 mm
Katalog # 761-0466
Marketsource
Leverandør: 3B Scientific
Gå til:
Product Details & Documents
These colourful anatomical charts depict various parts of the human body.
- Thick lamination ensures the chart does not curl up at the edges
- UV treatment ensures the chart does not get a faded yellow colour over time
- Printed on premium glossy UV resistant paper and thickly laminated on both sides
Leveringsinformation: Supplied with metal eyelets for easy hanging.
Specifikationer
- Beskrivelse:Chart, human skeleton, laminated
- B×H:500×670 mm