Milk butyrometer
Butyrometer for milk, 0 - 9% : 0,1
Katalog # GERB3155-C
Leverandør: GERBER FUNKE
Gå til:
Product Details & Documents
Borosilicate glass.
- With or without stopper
Available with a declaration of conformity per batch and type (in German only).
Specifikationer
- Beskrivelse:Butyrometer for milk, 0 - 9% : 0,1
- Seq:15
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Stopper, rubber, conical, for all measuring method butyrometersStopper, rubber, conical, for all measuring method butyrometers614-2028Log ind for tilgængelighed1 * 100 Items - 360,00 kr