Orders LinkContactUsLinkComponent
Cloning cylinders, PYREX®
Cloning cylinders, PYREX®
8 mm, 8 mm
Katalog # CORN3166-8
Leverandør:   Corning
Cloning cylinders, PYREX®
Katalog # CORN3166-8
Leverandør:  Corning
Cloning cylinders, PYREX®
Cloning cylinders, PYREX®
8 mm, 8 mm
Price
761,00 kr
Log ind for tilgængelighed
1 * 125 Items
Product Details & Documents
Cloning cylinders, made of borosilicate glass code 7740. Clone a single cell or group of cells by surrounding them with this open ended cloning cylinders. Dip the end of the cylinder into sterile silicone grease before pressing to the bottom of a culture dish to create an isolated well.
  • Reusable
  • Non sterile
  • Cylinders are autoclavable or can be dry heat sterilised
Documents
Specifikationer
  • Udv.Ø:
    8 mm
  • Højde:
    8 mm