Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for DOSE IT

Accessories for DOSE IT

Silicone tubing, autoclavable, length: 25 m (bulk roll), 4 mm
Katalog # INTG171034
Leverandør:   INTEGRA Biosciences
Accessories for DOSE IT
Katalog # INTG171034
Leverandør:  INTEGRA Biosciences
Accessories for DOSE IT
Accessories for DOSE IT

Silicone tubing, autoclavable, length: 25 m (bulk roll), 4 mm

Price
1.690,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Documents

Specifikationer
  • Beskrivelse:
    Silicone tubing, autoclavable, length: 25 m (bulk roll)
  • Ø indv.:
    4 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.