Thermal transfer printer labels, type B-424
Printer labels, B-424, Papir, Permanent, gummibaseret, Thermal transfer, hvid, 360X-Plus II, 76×50 mm, 1000 labels/roll
Katalog # BRDY030138
Leverandør: Brady
Gå til:
Product Details & Documents
Printable, paper labels for thermal transfer printers.
- Good contrast and smear resistance
Labels are ideally suited for use on smooth surfaces.
Specifikationer
- Printer:360X-Plus II
- Printing method:Thermal transfer
- Model:B-424
- Farve:hvid
- Beskrivelse:Printer labels
- Dimensioner:76×50 mm
- Materiale:Papir
- Seq:05
- Pakket:1000 labels/roll
- Klæbestof:Permanent, gummibaseret