Orders LinkContactUsLinkComponent
Sprøjtedispenser, Microlab® 600
Sprøjtedispenser, Microlab® 600
Dual syringe dispenser basic controller, Microlab 615-DIS, two 1 ml BFP default syringes
Catalog # 615-0025
Supplier:   HAMILTON BONADUZ
Sprøjtedispenser, Microlab® 600
Catalog # 615-0025
Supplier:  HAMILTON BONADUZ
Sprøjtedispenser, Microlab® 600
Sprøjtedispenser, Microlab® 600
Dual syringe dispenser basic controller, Microlab 615-DIS, two 1 ml BFP default syringes
Price
kr74,170.00
Log in for availability
1 * 1 Items
Product Details & Documents
The Microlab® 600 dispensers are highly precise syringe pumps with a touchscreen graphical user interface designed to quickly and easily dispense liquids. This positive displacement system provides better than 99% accuracy, independent of liquid viscosity, vapour pressure and temperature. The inert fluid path minimises sample carry over and improves compatibility with harsh chemicals. There are three different dispenser models available.
  • Dispenserer volumener fra 100 nl til 50 ml
  • Stabil væskevej, der kan anvendes til barske kemikalier
The single syringe dispenser is ideal for dedicated dispensing applications requiring a single syringe drive. The dual syringe dispenser has two syringe drives for applications such as animal dosing that require the use of more than one liquid at the same time. The dual syringe continuous dispenser is designed for time critical applications requiring repetitive dispensing.
The basic controller comes standard with a quick start run screen that provides basic diluting and dispensing functionality. The advanced controller delivers additional functions through intuitive wizards and the custom method editor. Wizard settings can be saved as favorites or custom methods on the hardware key. The basic controller can be upgraded to the advanced controller if it is not selected initially.
The standalone syringe pump offers total control with customised program methods using Visual C#®, Visual Basic®, and LabVIEW®. Commands can be deployed to any instrument on the network from anywhere in the world. The API is compatible with Windows® computers running Microsoft®. NET version 2.0 or higher.
Certifications: CE marked. CSA certified.
Accessories information: The Bubble Free Prime (BFP) syringe is specifically designed for the Microlab® 600 series and the only syringe type compatible with the instrument. It has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Ordering information: Dispensers are supplied with a Concorde hand probe (dual dispensers use the dual push button hand probe), universal valves, fill/dispense tubing, accessory holder, country specific power cord and default syringes. Standalone pumps are supplied with universal valves, country specific power cord and default syringes. To order a dispenser or standalone pump with other syringe sizes, please contact your local Avantor sales office.
Specifications
  • Beskrivelse:
    Dual syringe dispenser basic controller, Microlab 615-DIS, two 1 ml BFP default syringes
  • Seq:
    03
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI10150621
    Luer lock conversion kit
    Log in for availability
    1 * 1 Items - 996,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI1171-02
    Dispense tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEP
    Log in for availability
    1 * 1 Items - 648,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI1173-02
    Dispense tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEP
    Log in for availability
    1 * 1 Items - 648,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI64241-01
    Screen protector, PET
    Log in for availability
    1 * 2 Items - 498,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI61092-01
    Universal power supply, 110 - 220 VAC
    Log in for availability
    1 * 1 Items - 1.100,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI65160-01
    Cable management system for disposable tip hand probe
    Log in for availability
    1 * 1 Items - 1.390,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI68562-01
    Airshield for fresh air inside the instrument
    Log in for availability
    1 * 1 Items - 3.840,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1624
    Håndprobe med dobbelt trykknap
    Log in for availability
    1 * 1 Items - 7.300,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI1172-02
    Fill tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEP
    Log in for availability
    1 * 1 Items - 532,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI1174-02
    Fill tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEP
    Log in for availability
    1 * 1 Items - 532,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI61491-01
    Continuous fill tubing, volume 3,14 µl/mm, 12 gauge, FEP
    Log in for availability
    1 * 1 Items - 805,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI61500-05
    New Microlab 600 controller, includes SD card slot, serial port, large and small USB port
    Log in for availability
    1 * 1 Items - 24.320,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI61491-02
    Continuous fill tubing, volume 0,79 µl/mm, 18 gauge, FEP
    Log in for availability
    1 * 1 Items - 714,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-5822
    Printer kit includes printer, power source, adapter and cable
    Log in for availability
    1 * 1 Items
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI75702
    Disposable tips for 613-1625, 5 ml
    Log in for availability
    1 * 250 Items - 855,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1626
    Fodkontakt til ML 600-serien
    Log in for availability
    1 * 1 Items - 1.970,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI61729-01
    Valve plug
    Log in for availability
    1 * 1 Items - 183,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1625
    Håndprobe med engangsspids, stort volumen (1 - 5 ml)
    Log in for availability
    1 * 1 Items - 13.810,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI61710-01
    Probe holder and tubing stand
    Log in for availability
    1 * 1 Items - 1.350,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1582
    Fill tubing, volume 0,96 ml, 1219 mm, 18 gauge, FEP
    Log in for availability
    1 * 1 Items - 391,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1583
    Fill tubing, volume 4,57 ml, 1219 mm, 12 gauge, FEP
    Log in for availability
    1 * 1 Items - 459,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI61500-03
    Custom programmer kit (compatible with Microsoft® .NET 2.0 and higher)
    Log in for availability
    1 * 1 Items - 2.590,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1621
    Concorde CT håndprobe
    Log in for availability
    1 * 1 Items - 2.440,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI61498-01
    Valve cross tube assembly
    Log in for availability
    1 * 1 Items - 341,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1620
    Venstremontering
    Log in for availability
    1 * 1 Items - 2.950,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1584
    Dispense tubing, volume 1,08 ml, 1372 mm, 18 gauge, FEP
    Log in for availability
    1 * 1 Items - 399,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1585
    Dispense tubing, volume 5,15 ml, 1372 mm, 6,35 - 711,2 mm UNF, 12 gauge, FEP
    Log in for availability
    1 * 1 Items - 488,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1619
    Højremontering
    Log in for availability
    1 * 1 Items - 2.950,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-5823
    Disposable tip hand probe, 0,5 - 1000 µl
    Log in for availability
    1 * 1 Items - 6.620,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI88990
    Tubing clips
    Log in for availability
    1 * 5 Items - 523,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HAMI61500-02
    Basic to Advanced Controller upgrade kit, including 2 GB SD card
    Log in for availability
    1 * 1 Items - 6.440,00 kr