Wooden applicators
One pointed end applicators for cotton wool, Hårdttræ, 150 mm
Katalog # HECH44301015
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Wooden applicators of hardwood in bundles of 50 or 100 pieces.
- Unpointed (blunt), one end pointed and cotton ended variants are available
Ordering information: Unpointed (blunt), one end pointed and cotton ended variants.
Caution: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Beskrivelse:One pointed end applicators for cotton wool
- Længde:150 mm
- Materiale:Hårdttræ
- Seq:03