Orders LinkContactUsLinkComponent
Filter Holder Accessories
Hose connector 6,35 mm NPTF quick-release nipple,
stainless steel, PL-1 Cartridge Housings
Katalog # MILFXX6700028
Leverandør:   Merck
Filter Holder Accessories
Katalog # MILFXX6700028
Leverandør:  Merck
Filter Holder Accessories
Filter Holder Accessories
Hose connector 6,35 mm NPTF quick-release nipple,
stainless steel, PL-1 Cartridge Housings
Price
592,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Til:
    PL-1 Cartridge Housings
  • Beskrivelse:
    Hose connector 6,35 mm NPTF quick-release nipple,
    stainless steel
  • Seq:
    16