Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Filter Holder Accessories

Tubing for vacuum, 3,175 mm, fluoroelastomer, 190 mm
Katalog # MILFXX6504710
Leverandør:   Merck
Filter Holder Accessories
Katalog # MILFXX6504710
Leverandør:  Merck
Filter Holder Accessories
Filter Holder Accessories

Tubing for vacuum, 3,175 mm, fluoroelastomer, 190 mm

Price
405,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Tubing for vacuum, 3,175 mm, fluoroelastomer, 190 mm
  • Seq:
    17