Orders LinkContactUsLinkComponent
Jacalin for IgA purification
Jacalin for IgA purification
Jacalin, immobilised, Liquid, 2 mL
Catalog # 786-167
Supplier:   G-Biosciences
Jacalin for IgA purification
Catalog # 786-167
Supplier:  G-Biosciences
CAS Number:  
Jacalin for IgA purification
Jacalin for IgA purification
Jacalin, immobilised, Liquid, 2 mL
CAS Number:  
Price
2.690,00 kr
Log in for availability
1 * 2 mL
Product Details & Documents
Jacalin, or Artocarpus integrifolia lectin, is a tetrameric two-chain lectin with a molecular weight of 66 kDa. It is a α-D-galactose binding lectin purified from jack fruit (Artocarpus integrifolia) seeds.
Applications include isolating IgA from human serum and colostrums, isolating human plasma glycoproteins and histochemistry. It also binds IgD.
Specifications
Specifications
  • Pakn:
    2 mL
  • Beskrivelse:
    Jacalin, immobilised
  • Model:
    Liquid