Orders LinkContactUsLinkComponent
D-(+)-Cellobiose, Millipore®
D-(+)-Cellobiose, Millipore®
Glass bottle for solids, 1 kg
Katalog # 1.02352.1000
Leverandør:   Merck
D-(+)-Cellobiose, Millipore®
Katalog # 1.02352.1000
Leverandør:  Merck
CAS nummer:  528-50-7
D-(+)-Cellobiose, Millipore®
Glass bottle for solids, 1 kg
CAS nummer:  528-50-7
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
  • CAS nummer:528-50-7
  • Formel:C₁₂H₂₂O₁₁
  • MDL nummer:MFCD00136034
  • Molecular Weight:342.30 g/mol
  •  :M-Clarity™ quality level = MQ100
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-436-5
Documents
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg