Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Wipe rolls, Multiclean®

Wipe rolls, Multiclean®

Roll with embossing, 2 ply, blå, 350×370 mm, 2 rolls of 1000 sheets (360 m per roll)
Katalog # ZVGA11436-12
Wipe rolls, Multiclean®
Katalog # ZVGA11436-12
Wipe rolls, Multiclean®
Wipe rolls, Multiclean®

Roll with embossing, 2 ply, blå, 350×370 mm, 2 rolls of 1000 sheets (360 m per roll)

Price
977,00 kr
Log ind for tilgængelighed
1 * 2 ROL
Product Details & Documents

These perforated paper towel rolls are suitable for the removal of major dirt on surfaces. They are suitable for industry and trade applications, e.g. machine maintenance.

  • High absorbency and wet strength
  • 2 or 3 ply
  • Can be used with floor-standing and wall-mountable dispensers

Colour: Blue.

Specifikationer
  • Pakket:
    2 rolls of 1000 sheets (360 m per roll)
  • Farve:
    blå
  • Beskrivelse:
    Roll with embossing, 2 ply
  • L×B:
    350×370 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.