Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Natriumchlorid 0,9%, Ecotainer®

Bottles of 1000 ml, 6
  BRAU3570160
 :   B. Braun
Natriumchlorid 0,9%, Ecotainer®
  BRAU3570160
 :  B. Braun
 :  7647-14-5
Natriumchlorid 0,9%, Ecotainer®
Natriumchlorid 0,9%, Ecotainer®

Bottles of 1000 ml, 6

 :  7647-14-5
 
222,00 kr
   
1 * 6 Items
 

Sterile and pyrogen-free topical rinse solution.

  • PVC- and latex-free
  • Sterile and non-pyrogenic
  • Handy container with screw cap

Ecotainer® has been designed for intra- and post-operative irrigation.

 :  Saline, NaCl, Sea salt, Halite
  •  :7647-14-5
  •  :NaCl
  •  :MFCD00003477
  •  :58.44 g/mol

 

 

 

  • Pakn:
    6
  • Pk Info:
    Bottles of 1000 ml