Multi-panel drug tests
Product Details & Documents
These multipanel drugs tests can detect up to 12 drugs simultaneously.
- Clear results in less than three minutes
- Great accuracy, wide range of tests available
- For detecting recent drug use approximately 1 to 8 days
Suitable for on-site screening, safety critical situations, random screening, for cause screening, and monitoring rehabilitation.
AMP: Amphetamines ; BAR: Barbiturates ; BUP: Buprenorphine ; BZO: Benzodiazepines ; CLON: Clonezepam ; COC: Cocaine ; EDDP: Methadone Metabolite ; FEN: Fentanyl ; KET: Ketamine ; MAMP: Methamphetamines ; MDMA: Ecstasy ; MOP: Morphine ; MTD: Methadone ; OPI: Opiates ; OXY: Oxycodone ; PCP: Phencyclidine ; PPX: Propoxyphene ; TCA: Tricyclic Antidepressants ; TRA: Tramadol ; THC: Cannabis.
: ISO 9001, ISO 13485. IVD in accordance with 98/79/EC.
: Individually wrapped in a foil pouch, which ensures that the high-quality of the product is retained throughout the shelf-life.
Specifikationer
- Beskrivelse:Multi-panel drug tests
- Seq:035
- Analyser:(AMP300/mAMP300/BAR/BZO/COC150/KET/MDMA300/MOP/PCP/THC)