Ribbons for label printer BBP™11
Thermal transfer printer ribbon, 7964 series, width 110 mm, BBP™11, 70 m, sort
Katalog # BRDY804452
Leverandør: Brady
Gå til:
Product Details & Documents
Documents
Specifikationer
- Farve:sort
- Beskrivelse:Thermal transfer printer ribbon, 7964 series, width 110 mm
- Længde:70 m
- Seq:11
- Til:BBP™11