Ceramic beads, SiLibeads®
Ceramic beads, 2,60 - 3,30 mm, Zirconiumoxid
Katalog # LINR9729
Leverandør: SIGMUND LINDNER
Gå til:
Product Details & Documents
Ceramic beads made of Zirconium Oxide / Yttrium stabilised.
- High density
- Very high wear and tear resistance
- Low contamination of the grinding product
- Useable for all modern types of mills and high energy mills (vertical and horizontal)
- Excellent crystal structure avoids bead breakage and reduces the abrasion of mill parts
- No radioactivity
- Conformity to 1935/2004/EC
The exact roundness as well as the smooth and polished surface of the beads contributes to low wear rate.
Specifikationer
- Ø:2,60 - 3,30 mm
- Beskrivelse:Ceramic beads
- Materiale:Zirconiumoxid