Orders LinkContactUsLinkComponent
Accessories for comparators
Accessories for comparators
Manganese, 3/169, 0-4,0 mg/l
Catalog # LOVI230690
Supplier:   Lovibond
Accessories for comparators
Catalog # LOVI230690
Supplier:  Lovibond
Accessories for comparators
Accessories for comparators
Manganese, 3/169, 0-4,0 mg/l
Price
2.960,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Specifications
  • Område:
    0-4,0 mg/l
  • Seq:
    38
  • Kode:
    3/169
  • Testmetode:
    Manganese