Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Hardness quality control standard

Hardness quality control standard

Hardness quality control standard, low range, 100 mg/l as CaCO3 total hardness
50 mg/l as CaCO3 calcium hardness, 500 mL
Katalog # HACH2833449
Leverandør:   Hach
Hardness quality control standard
Katalog # HACH2833449
Leverandør:  Hach
CAS nummer:  
Hardness quality control standard
Hardness quality control standard

Hardness quality control standard, low range, 100 mg/l as CaCO3 total hardness
50 mg/l as CaCO3 calcium hardness, 500 mL

CAS nummer:  
Price
884,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

These mixed standards are ready to use with no dilutions required. Includes standards for both total hardness and calcium hardness determination.

Documents

Specifikationer

Specifikationer

  • Koncentration:
    100 mg/l as CaCO<sub>3</sub> total hardness<br />50 mg/l as CaCO<sub>3</sub> calcium hardness
  • Pakn:
    500 mL
  • Beskrivelse:
    Hardness quality control standard, low range

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.