Pasteur pipettes
—, 150 mm, —, +, Individually wrapped
Katalog # KART329
Leverandør: KARTELL
Gå til:
Product Details & Documents
PE, disposable, sterile or non sterile.
- Disposable PE pipettes with bulb and graduations
- The slender tip guarantees precision transfer of the sample
- Non sterile pipettes are available in multi-packs; sterile pipettes are individually wrapped
Caution: *Capillary
Specifikationer
- Pakket:Individually wrapped
- Steril:+
- Kapacitet:—
- Længde:150 mm
- Seq:1
- Inddeling:—