Height adjustable worktable
Concept motor table, (motorised height adjustment) ; control panel with three memory settings, 1500×750×700/1100 mm
Katalog # SOVEC10341037
Leverandør: TRESTON
Gå til:
Product Details & Documents
These worktables are a suitable solution for industrial laboratories and research and development centers. They are designed to accommodate the necessary measurement equipment and other tools ergonomically at a safe distance for use. These laboratory benches fit also to education purposes. Modularity and accessories are individually adjustable according the task and human requirements.
- Robust, elegant, and very flexible
- Easy to adapt to individual needs, new equipment, and changes in the working environment
- With durable table frame
- Leveling glides
Ordering information: Table top sold separately; contact your Avantor representative for more information.
Specifikationer
- Beskrivelse:Concept motor table, (motorised height adjustment) ; control panel with three memory settings
- B×D×H:1500×750×700/1100 mm