Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Drawer cabinets 75/56

Drawer cabinets 75/56

Cabinet with lock, 2 doors, 750×520×500 mm
Katalog # SOVE60849106
Leverandør:   TRESTON
Drawer cabinets 75/56
Katalog # SOVE60849106
Leverandør:  TRESTON
Drawer cabinets 75/56

Cabinet with lock, 2 doors, 750×520×500 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Drawer units 75 series designed for storing lightweight items, in environments such as light assembly, inspection, or testing laboratory. These units equipped with base for stand alone version and casters for mobile version.

  • Castors equipped with brakes
  • Maximum loading capacity: 60 kg per drawer

Colour: grey

Specifikationer
  • Beskrivelse:
    Cabinet with lock
  • Seq:
    02
  • B×D×H:
    750×520×500 mm
  • Døre:
    2 doors
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.