Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Drenchers

Drenchers

Drencher, bent, 25 mm, 0,9 mm
Katalog # SOCO376.0925B
Leverandør:   Socorex Isba
Drenchers
Katalog # SOCO376.0925B
Leverandør:  Socorex Isba
Drenchers
Drenchers

Drencher, bent, 25 mm, 0,9 mm

Price
225,00 kr
Log ind for tilgængelighed
1 * 3 Items
Product Details & Documents

These straight or bent drenchers are made from stainless steel. They are ideal for safe animal nose and mouth feeding.

  • Luer lock fittings
Specifikationer
  • Udv.Ø:
    0,9 mm
  • Beskrivelse:
    Drencher, bent
  • Længde:
    25 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.