Orders LinkContactUsLinkComponent
Supelco® COD CRM, standard solutions
Supelco® COD CRM, standard solutions
COD standard solution, 1000 mg/L, ± 11 mg/L , 1.14541; 1.14691; 1.14555, Plastic bottle, 3 - 5, 100 mL
Katalog # 1.25032.0100
Leverandør:   Merck
Supelco® COD CRM, standard solutions
Katalog # 1.25032.0100
Leverandør:  Merck
CAS nummer:  
Supelco® COD CRM, standard solutions
Supelco® COD CRM, standard solutions
COD standard solution, 1000 mg/L, ± 11 mg/L , 1.14541; 1.14691; 1.14555, Plastic bottle, 3 - 5, 100 mL
CAS nummer:  
Price
383,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Certifications: Traceable to SRM from NIST and USP.
Specifikationer
Specifikationer
  • Koncentration:
    1000 mg/L
  • Pakning:
    Plastic bottle
  • Pakn:
    100 mL
  • Beskrivelse:
    COD standard solution
  • Nøjagtighed:
    ± 11 mg/L
  • pH-område:
    3 - 5
  • För testkit(s):
    1.14541; 1.14691; 1.14555
  • Cat. No.:
    1.25032.0100
  • M-Clarity quality level:
    MQ200