Sample cups for analysers, ratiolab®
For Technicon T1 analysers, 2 ml
Katalog # RATI5700110
Leverandør: RATIOLAB
Gå til:
Product Details & Documents
Made of clear PS.
- Good chemical resistance
- Good temperature resistance
Analysis beakers offer the highest precision for use with robotic analytical systems.
Specifikationer
- Kapacitet:2 ml
- Beskrivelse:For Technicon T1 analysers
- Seq:02