Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

100 ml, 0,080 ± ml, gul
Katalog # KAVA632433111030
Leverandør:   Kavalier
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Katalog # KAVA632433111030
Leverandør:  Kavalier
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

100 ml, 0,080 ± ml, gul

Price
488,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Soda-lime glass.

  • Enlarged form
  • Non autoclavable at 121 °C

 : DIN 12691, ISO 648

Specifikationer
  • Farvekode:
    gul
  • Kapacitet:
    100 ml
  • Seq:
    12
  • Tolerance:
    0,080 ± ml