Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

5 ml, 0,015 ± ml, hvid
Katalog # KAVA632433111018
Leverandør:   Kavalier
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Katalog # KAVA632433111018
Leverandør:  Kavalier
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

5 ml, 0,015 ± ml, hvid

Price
76,60 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Soda-lime glass.

  • Enlarged form
  • Non autoclavable at 121 °C

Certifications: DIN 12691, ISO 648

Specifikationer
  • Farvekode:
    hvid
  • Kapacitet:
    5 ml
  • Seq:
    03
  • Tolerance:
    0,015 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.