Orders LinkContactUsLinkComponent
Silicone rubber stoppers, conical
Silicone rubber stoppers, conical
2, 35,0 mm, 7 mm, 31,0 mm, 38,0 mm
Katalog # DENE1012531
Leverandør:   DEUTSCH NEUMANN
Silicone rubber stoppers, conical
Katalog # DENE1012531
Leverandør:  DEUTSCH NEUMANN
Silicone rubber stoppers, conical
Silicone rubber stoppers, conical
2, 35,0 mm, 7 mm, 31,0 mm, 38,0 mm
Price
70,40 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These stoppers are biologically inert and transparent.
  • Resistant to temperatures from −60 to +220 °C and for short periods up to +250 °C
Certifications: DIN 12 871
Specifikationer
  • Ø bund:
    31,0 mm
  • Ø top:
    38,0 mm
  • Hul Ø:
    7 mm
  • Ant. huller:
    2
  • Højde:
    35,0 mm