Orders LinkContactUsLinkComponent
Thread adapters for pumps
Thread adapters for pumps
Adapter set, contains one each of thread adapter Tri-Sure®/50,8 mm BSP, Mauser/50,8 mm BSP, DIN 61 - 50,8 mm BSP and DIN 71 - 50,8 mm BSP
Katalog # BURK0600-3010
Leverandør:   BURKLE
Thread adapters for pumps
Katalog # BURK0600-3010
Leverandør:  BURKLE
Thread adapters for pumps
Thread adapters for pumps
Adapter set, contains one each of thread adapter Tri-Sure®/50,8 mm BSP, Mauser/50,8 mm BSP, DIN 61 - 50,8 mm BSP and DIN 71 - 50,8 mm BSP
Price
657,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Adapter set, contains one each of thread adapter Tri-Sure®/50,8 mm BSP, Mauser/50,8 mm BSP, DIN 61 - 50,8 mm BSP and DIN 71 - 50,8 mm BSP
  • Seq:
    01