Chemically resistant diaphragm dosing pumps SIMDOS®
SIMDOS FEM 1.10FT.18 S dosing pump, 144 mm, 150 mm, FT in PTFE (polytetrafluoroethylene), 93 mm
Katalog # 224-0065
Leverandør: KNF
Gå til:
Product Details & Documents
Simple, user-friendly compact dosing pump.
Application: Metering liquids.
- Large screen and intuitive interface with drop down menus, easy calibration
- Single programming button
- Adaptable to fluid characteristic
- Pump head located externally for better protection of electronic components
- Self-priming up to 3 mCE, can run dry
- Dosage of 1 to 100 ml/min
- Dose reproducibility ±1%
- Simplified dose programming from 1 to 1000 ml
- Dispensed volume 1 to 1000 ml
- Maximum pressure: 6 bar
- Maximum viscosity: 150 cS
- Flow rate from 0,03 up to 100 ml/min
- Pressure head 60 mWg, suction head 2 mWg and 3 mWg respectively
- Easy exchange of the transfer diaphragm by activating the maintenance command in the operating program
- Small footprint
Pump heads available in PP, PVDF or PTFE - diaphragms available in PTFE, valves in FFKM.
Flow rate can either be set manually (version S) or both manually and via an external control device as well as with interface RC232 (version RCP)
S version:
Manual control by rotary knob
RC version:
Controlled by rotary knob
Analogue control (0 to 10 V, 0 to 100 mA, 4 to 20 mA from 0 to 100%)
Start/stop by logical control (TTL); reset/prime by logical control (TTL)
Alarm output signal
Cable for external control included; control pedal available as an option
Bestillingsoplysninger: Available with pump head in different materials.
Specifikationer
- Beskrivelse:SIMDOS FEM 1.10FT.18 S dosing pump
- Højde:144 mm
- Længde:150 mm
- Pumpehoved:FT in PTFE (polytetrafluoroethylene)
- Bredde:93 mm
- Doseringsvolumen:30 µl - 999 ml
- Cat. No.:224-0065
- Gentagefunktion:±1 %
- Tilladt væsketemperatur:5...80 °C
- Trykhøjde:60 mWs
- Vægt:0,9 kg
- Tilladt omgivende temperatur:5...40 °C
- Model:S-Version
- Flowhastighed:20
- Maks. tryk:6 bar
- Forbindelse:Hydraulic connection UNF 1/4"-28
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Column fixture
Produktinformation
- Katalog #TilgængelighedPrice
- Column fixture
Produktinformation
- Katalog #TilgængelighedPrice
- In-line filter FS 25 T PVDF mesh opening 70 µm SIMDOS® 10In-line filter FS 25 T PVDF mesh opening 70 µm SIMDOS® 10KNFA165211Log ind for tilgængelighed1 * 1 Items - 654,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- SIMDOS spare parts kit, valves and seals, all versionsSIMDOS spare parts kit, valves and seals, all versionsKNFA160186Log ind for tilgængelighed1 * 1 Items - 2.230,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- In-line filter FS 60 X PEEK mesh opening, UNF ¼"-28 connectorIn-line filter FS 60 X PEEK mesh opening, UNF ¼"-28 connectorKNFA323625Log ind for tilgængelighed1 * 1 Items - 826,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Kit pressure control valve JFDV 30 KTZ for FEM 1,02 KTKit pressure control valve JFDV 30 KTZ for FEM 1,02 KTKNFA166286Log ind for tilgængelighed1 * 1 Items - 3.520,00 kr