Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Basic Violet 3, Sigma-Aldrich®

Basic Violet 3, Sigma-Aldrich®

Glass bottle for solids, 500 g
Katalog # 48770-500G
Ætsende
Sundhed
Irriterende
Miljø
Leverandør:   SIGMA ALDRICH MICROSCOPY
Basic Violet 3, Sigma-Aldrich®
Katalog # 48770-500G
CAS nummer:  548-62-9
Basic Violet 3, Sigma-Aldrich®
Basic Violet 3, Sigma-Aldrich®

Glass bottle for solids, 500 g

CAS nummer:  548-62-9
Price
2.450,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Mixture of crystal violet [C.I.No. 42555] and methyl violet [C.I.No. 42535]. Dye employed for preparing Gram stain.

Synonymer:  4-[4,4'-Bis(dimethylamino)benzhydryliden]cyclohexa-2,5-dien-1-yliden]dimethylammoniumchlorid, CI 42555
  • CAS nummer:548-62-9
  • Formel:C₂₅H₃₀ClN₃
  • MDL nummer:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Merck Index:13,04406
  • Smeltende Pt:189…194 °C
  • Tæthed:1,19 g/cm³ (20 °C)
  • Opbevaringstemperatur:Room Temperature
  • EINECS:208-953-6
  • FN:3077
  • ADR:9,III

Documents

Specifikationer

Specifikationstestresultater

  • Appearance (colour):
    Green to very dark green
  • Appearance (form):
    Powder
  • Thin layer chromatogr.:
    Corresponds to standard
  • Solubility (colour):
    0.01 g in 10 ml methanol
  • Solubility (method):
    0.01 g in 10 ml methanol
  • Loss on drying:
    ≤10%
  • Wavelength (1) (UV):
    580 - 595 nm
  • Extinction (1%/1 cm) (1) (UV):
    ≥1300
  • Solvent (UV):
    Water

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.