Basic Violet 3, Sigma-Aldrich®
25 g
Katalog # 48770-25G
Leverandør: SIGMA ALDRICH MICROSCOPY
Basic Violet 3, Sigma-Aldrich®
Katalog # 48770-25G
Leverandør: SIGMA ALDRICH MICROSCOPY
CAS nummer: 548-62-9
Gå til:
Product Details & Documents
Mixture of crystal violet [C.I.No. 42555] and methyl violet [C.I.No. 42535]. Dye employed for preparing Gram stain.
Synonymer: 4-[4,4'-Bis(dimethylamino)benzhydryliden]cyclohexa-2,5-dien-1-yliden]dimethylammoniumchlorid, CI 42555
- CAS nummer:548-62-9
- Formel:C₂₅H₃₀ClN₃
- MDL nummer:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck Index:13,04406
- Smeltende Pt:189…194 °C
- Tæthed:1,19 g/cm³ (20 °C)
- Opbevaringstemperatur:Room Temperature
- EINECS:208-953-6
- FN:3077
- ADR:9,III
Documents
Merck The Color of Tissue Diagnostics
Produktlitteratur
Specifikationer
Specifikationstestresultater
- Appearance (colour):Green to very dark green
- Appearance (form):Powder
- Thin layer chromatogr.:Corresponds to standard
- Solubility (colour):0.01 g in 10 ml methanol
- Solubility (method):0.01 g in 10 ml methanol
- Loss on drying:≤10%
- Wavelength (1) (UV):580 - 595 nm
- Extinction (1%/1 cm) (1) (UV):≥1300
- Solvent (UV):Water
Specifikationer
- Pakn:25 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



