Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Basic Violet 3, Sigma-Aldrich®

Glass bottle for solids, 100 g
Katalog # 48770-100G
Ætsende
Sundhed
Irriterende
Miljø
Leverandør:   SIGMA ALDRICH MICROSCOPY
Basic Violet 3, Sigma-Aldrich®
Katalog # 48770-100G
CAS nummer:  548-62-9
Basic Violet 3, Sigma-Aldrich®
Basic Violet 3, Sigma-Aldrich®

Glass bottle for solids, 100 g

CAS nummer:  548-62-9
Price
744,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Mixture of crystal violet [C.I.No. 42555] and methyl violet [C.I.No. 42535]. Dye employed for preparing Gram stain.

Synonymer:  4-[4,4'-Bis(dimethylamino)benzhydryliden]cyclohexa-2,5-dien-1-yliden]dimethylammoniumchlorid, CI 42555
  • CAS nummer:548-62-9
  • Formel:C₂₅H₃₀ClN₃
  • MDL nummer:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Merck Index:13,04406
  • Smeltende Pt:189…194 °C
  • Tæthed:1,19 g/cm³ (20 °C)
  • Opbevaringstemperatur:Room Temperature
  • EINECS:208-953-6
  • FN:3077
  • ADR:9,III
Specifikationer

Specifikationstestresultater

  • Appearance (colour):
    Green to very dark green
  • Appearance (form):
    Powder
  • Thin layer chromatogr.:
    Corresponds to standard
  • Solubility (colour):
    0.01 g in 10 ml methanol
  • Solubility (method):
    0.01 g in 10 ml methanol
  • Loss on drying:
    ≤10%
  • Wavelength (1) (UV):
    580 - 595 nm
  • Extinction (1%/1 cm) (1) (UV):
    ≥1300
  • Solvent (UV):
    Water

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer