Basic Violet 3, Sigma-Aldrich®
Glass bottle for solids, 25 g
Katalog # C0775-25G
Leverandør: SIGMA ALDRICH MICROSCOPY
Basic Violet 3, Sigma-Aldrich®
Katalog # C0775-25G
Leverandør: SIGMA ALDRICH MICROSCOPY
CAS nummer: 548-62-9
Gå til:
Product Details & Documents
Synonymer: 4-[4,4'-Bis(dimethylamino)benzhydryliden]cyclohexa-2,5-dien-1-yliden]dimethylammoniumchlorid, CI 42555
- CAS nummer:548-62-9
- Formel:C₂₅H₃₀ClN₃
- MDL nummer:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck Index:13,04406
- Smeltende Pt:189…194 °C
- Tæthed:1,19 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:208-953-6
- FN:3077
- ADR:9,III
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g



