Supelco® Organic Reference Standard, Digoxin
Digoxin, Analytical standard, 5 g
D6003-5G
: Merck
- GHS signal: GHS06
: P260 - P264 - P270 - P301 + P310 - P304 + P340 + P311 - P314
- Pakn:5 g
- Standard:Analytical standard
- Beskrivelse:Digoxin
- Cat. No.:D6003-5G
- M-Clarity quality level:MQ100
- CAS-nr:20830-75-5
- Molekylvægt:780,94
- MDL No.:MFCD00003674
- Fareklassebeskrivelse:H300 - H331 - H373
- Opbevaring betingelser:RT
- Synonym(s):12β-Hydroxydigitoxin
- Shipping conditions:STD
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



