Supelco® Organic Reference Standard, Digoxin
Digoxin, Analytical standard, 1 g
Katalog # D6003-1G
Leverandør: Merck
Gå til:
Product Details & Documents
- GHS signal: GHS06
: P260 - P264 - P270 - P301 + P310 - P304 + P340 + P311 - P314
Specifikationer
Specifikationer
- Pakn:1 g
- Standard:Analytical standard
- Beskrivelse:Digoxin
- Cat. No.:D6003-1G
- M-Clarity quality level:MQ100
- CAS-nr:20830-75-5
- Molekylvægt:780,94
- MDL No.:MFCD00003674
- Fareklassebeskrivelse:H300 - H331 - H373
- Opbevaring betingelser:RT
- Synonym(s):12β-Hydroxydigitoxin
- Shipping conditions:STD



