Supelco® Organic Reference Standard, Digoxin
Digoxin, Analytical standard, 100 mg
Catalog # D6003-100MG
Supplier: Merck
Jump to:
Product Details & Documents
- GHS signal: GHS06
Caution: P260 - P264 - P270 - P301 + P310 - P304 + P340 + P311 - P314
Specifications
Specifications
- Pakn:100 mg
- Standard:Analytical standard
- Beskrivelse:Digoxin
- Cat. No.:D6003-100MG
- M-Clarity quality level:MQ100
- CAS-nr:20830-75-5
- Molekylvægt:780,94
- MDL No.:MFCD00003674
- Fareklassebeskrivelse:H300 - H331 - H373
- Opbevaring betingelser:RT
- Synonym(s):12β-Hydroxydigitoxin
- Shipping conditions:STD
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



