Chloroform solution, NMR reference standard, Supelco®
Chloroform solution, 20% in acetone-d6 (99.9 atom % D), NMR reference standard, NMR tube size 5 mm x 8 in., 1
Katalog # 611859-1EA
Leverandør: Merck
Chloroform solution, NMR reference standard, Supelco®
Katalog # 611859-1EA
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Koncentration:20% in acetone-d6 (99.9 atom % D)
- Pakn:1
- Standard:NMR reference standard
- Beskrivelse:Chloroform solution
- Pk Info:NMR tube size 5 mm x 8 in.
- Cat. No.:611859-1EA
- Beskrivelse:Chloroform solution
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



