Staining jars
Farvningsbakke, rustfrit stål, - mm
Katalog # MARI6618000
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Staining set consists of glass dish and staining jar made of soda lime glass, and staining tray made of either pressed glass or stainless steel.
These staining tray hold up to 10 slides (76×26 mm).
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.
Specifikationer
- Beskrivelse:Farvningsbakke, rustfrit stål
- B×D×H:- mm