Orders LinkContactUsLinkComponent
Filling cannulae
Filling cannulae
Filling cannulae, G 20, 100 mm
Katalog # MURRM720/06
Leverandør:   MURRAY S
Filling cannulae
Katalog # MURRM720/06
Leverandør:  MURRAY S
Filling cannulae
Filling cannulae, G 20, 100 mm
Price
374,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
Bestillingsoplysninger: Different sizes and gauges are available.
Documents
Specifikationer
  • Måler:
    G 20
  • Beskrivelse:
    Filling cannulae
  • Længde:
    100 mm
  • Seq:
    015