Hypodermic needles
Hypodermic, sharp, 18, 150 mm
Katalog # MURRM618/08
Leverandør: MURRAY S
Gå til:
Product Details & Documents
Non sterile needles with sharp ends and luer lock fitting.
Specifikationer
- Måler:18
- Beskrivelse:Hypodermic, sharp
- Længde:150 mm
- Seq:16