n-Hexadecan 99%, pure
Glass bottle, 2,5 L
Leverandør: Thermo Fisher Scientific
n-Hexadecan 99%, pure
Katalog # ACRO120460025
Leverandør: Thermo Fisher Scientific
CAS nummer: 544-76-3
Gå til:
Product Details & Documents
Synonymer: Hexadecane, Cetane
- CAS nummer:544-76-3
- Formel:H₃C(CH₂)₁₄CH₃
- MDL nummer:MFCD00008998
- Molecular Weight:226.45 g/mol
- Kogende Pt:287 °C (760 mmHg)
- Smeltende Pt:18 °C
- Tæthed:0,773 g/cm³
- Flash Pt:135 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:208-878-9
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid or solid
- Infrared spectrum:Conforms
- Refractive index:1.4330 to 1.4350 (20°C, 589 nm)
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



